Learn More
Med Vet International McKesson Beige Cotton Tubular Stockinette, 8 Inch x 25 Yard, 16-4T-825, Each
Catalog No. 503043421
Encompass_Preferred
Stockinette Tubular McKesson 8 Inch X 25 Yard Cotton NonSterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Designed for use with synthetic casting systems because of wicking action and fast-drying features Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Cotton Beige 8 inches x 25 yards
Catalog No. 50-304-3421 Supplier Med Vet International Supplier No. 457633RL
Stockinette Tubular McKesson 8 Inch X 25 Yard Cotton NonSterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Designed for use with synthetic casting systems because of wicking action and fast-drying features Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Cotton Beige 8 inches x 25 yards