Learn More
Med Vet International McKesson White Polyester Tubular Stockinette, 4 Inch x 25 Yard, 16-4TS-425, Each
Catalog No. 503043398
Encompass_Preferred
Stockinette Tubular McKesson 4 Inch X 25 Yard Polyester NonSterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Designed for use with synthetic casting systems because of wicking action and fast-drying features Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Polyester White 4 inches x 25 yards
Catalog No. 50-304-3398 Supplier Med Vet International Supplier No. 457644RL
Stockinette Tubular McKesson 4 Inch X 25 Yard Polyester NonSterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Designed for use with synthetic casting systems because of wicking action and fast-drying features Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Polyester White 4 inches x 25 yards