Learn More
Hampton Research STOCKOPT ADA #01 PH 5.6
SDP
Catalog No. 50563239
Encompass
HR2-255-01/StockOptions ADA #01, 1.0 M ADA pH 5.6, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-239 Supplier Hampton Research Supplier No. HR225501
HR2-255-01/StockOptions ADA #01, 1.0 M ADA pH 5.6, 185 ml. Please note items from this supplier are non-returnable.