Learn More
Hampton Research STOCKOPT ADA #16 PH 7.1
SDP
Catalog No. 50563254
Encompass
HR2-255-16/StockOptions ADA #16, 1.0 M ADA pH 7.1, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-254 Supplier Hampton Research Supplier No. HR225516
HR2-255-16/StockOptions ADA #16, 1.0 M ADA pH 7.1, 185 ml. Please note items from this supplier are non-returnable.