Learn More
Hampton Research STOCKOPT GLYCINE #09 PH 9.4
SDP
Catalog No. 50563155
Encompass
HR2-250-09/StockOptions Glycine #09, 1.0 M Glycine pH 9.4, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-155 Supplier Hampton Research Supplier No. HR225009
HR2-250-09/StockOptions Glycine #09, 1.0 M Glycine pH 9.4, 185 ml. Please note items from this supplier are non-returnable.