Learn More
Hampton Research STOCKOPT GLYCINE #15 PH 10.0
SDP
Catalog No. 50563161
Encompass
HR2-250-15/StockOptions Glycine #15, 1.0 M Glycine pH 10.0, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-161 Supplier Hampton Research Supplier No. HR225015
HR2-250-15/StockOptions Glycine #15, 1.0 M Glycine pH 10.0, 185 ml. Please note items from this supplier are non-returnable.