Learn More
Hampton Research STOCKOPT GLYCINE #16 PH 10.1
SDP
Catalog No. 50563162
Encompass
HR2-250-16/StockOptions Glycine #16, 1.0 M Glycine pH 10.1, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-162 Supplier Hampton Research Supplier No. HR225016
HR2-250-16/StockOptions Glycine #16, 1.0 M Glycine pH 10.1, 185 ml. Please note items from this supplier are non-returnable.