Learn More
Hampton Research STOCKOPT GLYCINE #19 PH 10.4
SDP
Catalog No. 50563165
Encompass
HR2-250-19/StockOptions Glycine #19, 1.0 M Glycine pH 10.4, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-165 Supplier Hampton Research Supplier No. HR225019
HR2-250-19/StockOptions Glycine #19, 1.0 M Glycine pH 10.4, 185 ml. Please note items from this supplier are non-returnable.