Learn More
Hampton Research STOCKOPT GLYCINE #21 PH 10.6
SDP
Catalog No. 50563167
Encompass
HR2-250-21/StockOptions Glycine #21, 1.0 M Glycine pH 10.6, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-167 Supplier Hampton Research Supplier No. HR225021
HR2-250-21/StockOptions Glycine #21, 1.0 M Glycine pH 10.6, 185 ml. Please note items from this supplier are non-returnable.