Learn More
Hampton Research STOCKOPT MALIC ACID #03 PH 3.9
SDP
Catalog No. 50563100
Encompass
HR2-240-03/StockOptions Malic Acid #03, 1.0 M DL-Malic acid pH 3.9, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-100 Supplier Hampton Research Supplier No. HR224003
HR2-240-03/StockOptions Malic Acid #03, 1.0 M DL-Malic acid pH 3.9, 185 ml. Please note items from this supplier are non-returnable.