Learn More
Hampton Research STOCKOPT MALIC ACID #04 PH 4.0
SDP
Catalog No. 50563101
Encompass
HR2-240-04/StockOptions Malic Acid #04, 1.0 M DL-Malic acid pH 4.0, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-101 Supplier Hampton Research Supplier No. HR224004
HR2-240-04/StockOptions Malic Acid #04, 1.0 M DL-Malic acid pH 4.0, 185 ml. Please note items from this supplier are non-returnable.