Learn More
Hampton Research STOCKOPT MALIC ACID #09 PH 4.5
SDP
Catalog No. 50563106
Encompass
HR2-240-09/StockOptions Malic Acid #09, 1.0 M DL-Malic acid pH 4.5, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-106 Supplier Hampton Research Supplier No. HR224009
HR2-240-09/StockOptions Malic Acid #09, 1.0 M DL-Malic acid pH 4.5, 185 ml. Please note items from this supplier are non-returnable.