Learn More
Hampton Research STOCKOPT MALIC ACID #12 PH 4.8
SDP
Catalog No. 50563109
Encompass
HR2-240-12/StockOptions Malic Acid #12, 1.0 M DL-Malic acid pH 4.8, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-109 Supplier Hampton Research Supplier No. HR224012
HR2-240-12/StockOptions Malic Acid #12, 1.0 M DL-Malic acid pH 4.8, 185 ml. Please note items from this supplier are non-returnable.