Learn More
Hampton Research STOCKOPT MALIC ACID #13 PH 4.9
SDP
Catalog No. 50563110
Encompass
HR2-240-13/StockOptions Malic Acid #13, 1.0 M DL-Malic acid pH 4.9, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-110 Supplier Hampton Research Supplier No. HR224013
HR2-240-13/StockOptions Malic Acid #13, 1.0 M DL-Malic acid pH 4.9, 185 ml. Please note items from this supplier are non-returnable.