Learn More
Hampton Research STOCKOPT MOPS #07 PH 7.1
SDP
Catalog No. 50563192
Encompass
HR2-252-07/StockOptions MOPS #07, 1.0 M MOPS pH 7.1, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-192 Supplier Hampton Research Supplier No. HR225207
HR2-252-07/StockOptions MOPS #07, 1.0 M MOPS pH 7.1, 185 ml. Please note items from this supplier are non-returnable.