Learn More
Hampton Research STOCKOPT MOPS #13 PH 7.7
SDP
Catalog No. 50563198
Encompass
HR2-252-13/StockOptions MOPS #13, 1.0 M MOPS pH 7.7, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-198 Supplier Hampton Research Supplier No. HR225213
HR2-252-13/StockOptions MOPS #13, 1.0 M MOPS pH 7.7, 185 ml. Please note items from this supplier are non-returnable.