Learn More
Hampton Research HR2-102/StockOptions Hepes, pH 6.8 to 8.2 in 0.1 pH unit increments - 10 ml, tube format
SDP
Catalog No. 500936
Encompass
HR2-102/StockOptions Hepes, pH 6.8 to 8.2 in 0.1 pH unit increments - 10 ml, tube format. Please note items from this supplier are non-returnable.
Catalog No. 50-093-6 Supplier Hampton Research Supplier No. HR2102
HR2-102/StockOptions Hepes, pH 6.8 to 8.2 in 0.1 pH unit increments - 10 ml, tube format. Please note items from this supplier are non-returnable.