Learn More
Hampton Research HR2-249/StockOptions Succinic Acid, pH 4.3 - 6.6 in 0.1 pH unit increments - 10 ml, tube format
SDP
Catalog No. 50563122
Encompass
HR2-249/StockOptions Succinic Acid, pH 4.3 - 6.6 in 0.1 pH unit increments - 10 ml, tube format. Please note items from this supplier are non-returnable.
Catalog No. 50-563-122 Supplier Hampton Research Supplier No. HR2249
HR2-249/StockOptions Succinic Acid, pH 4.3 - 6.6 in 0.1 pH unit increments - 10 ml, tube format. Please note items from this supplier are non-returnable.