Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoko™ Kresto™ Classic Cleanser

Catalog No. 19048310
Encompass_Preferred
Change view
Click to view available options
Volume (English):
67.62 oz.
Packaging:
Bottle, one pump
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Packaging Quantity
19-048-310 67.62 oz. Bottle, one pump 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-048-310 Supplier Stoko™ Supplier No. PN98704406/DEL

missing translation for 'validMainframeMsg'

Quickly and effectively cleans grease, oil, grime, ink, carbon black, sewer sludge, adhesives, and other industrial dirt

  • Concentrated formula with ASTOPON™ natural walnut shell powder scrubber
  • Eucornol™ for extra skin protection
  • Gentle low-solvent formulation
  • Pleasant fragrance and pH balanced
  • Completely biodegradable
  • Will not clog drains or pipes

Specifications

Product Type Kresto™ Cleanser
Color Light Beige
Form Paste
Composition Water, C13-14 Isoparaffin, Walnut Shell Powder, Sodium Laureth Sulfate, Sulfated Castor Oil, Sodium Chloride, Cocamidopropyl Betaine, Quaternium 18 Bentonite, Cellulose Gum, Titanium Dioxide, Citric Acid, Fragrance, Potassium Sorbate, 2-Bromo-2-Nitropropane-1,3-Diol
For Use With (Application) Quickly and effectively cleans grease, oil, grime, ink, carbon black, sewer sludge, adhesives, and other industrial dirt
Packaging Bottle, one pump
pH Range 5.2 to 5.8
Quantity 1 Ea.
Volume (English) 67.62 oz.
Volume (Metric) 2000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.