Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoko™ Kwik-Wipes™ Towelettes

Catalog No. 19819788
Change view
Click to view available options
Quantity:
6 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-819-788 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-819-788 Supplier Stoko™ Supplier No. 2 87008 04

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perfect when a sink is unavailable or at individual work stations

  • Textured wipes for quick cleaning of dirt, grease, and grime from hands

Order Info

Convenience packaged 70 wipes per container, six containers per case; bulk packaged 130 wipes per container, four containers per case.

Specifications

Product Type Kwik-Wipes Towelette
Color White
Form Wipe
Composition Water, Dipropylene Glycol Methyl Ether, Sodium Caprylyl Sulfonate, Nonoxynol-10, Sodium Laureth Sulfate, PEG-75 Lanolin, fragrance, 2-Bromo-2-Nitropropane-1,3-Diol
For Use With (Application) For use on skin, also great for tools and other surfaces, quick cleaning of dirt, grease, and grime from hands
Packaging Canister
pH Range 6
Quantity 6 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.