Learn More
MSC SC Johnson Professional Skin Conditioning Cream, Fragrance-Free, Dye-Free, 1 L
Catalog No. 502868893
Encompass
Stokolan Light PURE is a lightly textured skin conditioning cream designed for professional use to help care for normal to dry skin. It is fragrance-free and dye-free. It is intended for use by those with moderate skin dryness from daily working in industrial, food handling, healthcare, office, and commercial areas.
  • Non-greasy formula for good skin condition
  • Skin hypoallergenic
  • Fragrance-free and dye-free
  • Contains Shea Butter to lock in hydration
  • Contains Allantoin for moisturization
  • Contains Glycerin for smooth skin
  • Silicone-free to prevent compatibility issues
  • Suitable for food facilities
  • Compatible with rubber production processes
  • Compatible with alcohol-based hand sanitizers
  • Proven glove compatibility
  • Chlorhexidine Gluconate (CHG) Compatible
  • Meets ECARF criteria for skin tolerability
Catalog No. 50-286-8893 Supplier MSC Supplier No. 62909718
Stokolan Light PURE is a lightly textured skin conditioning cream designed for professional use to help care for normal to dry skin. It is fragrance-free and dye-free. It is intended for use by those with moderate skin dryness from daily working in industrial, food handling, healthcare, office, and commercial areas.
  • Non-greasy formula for good skin condition
  • Skin hypoallergenic
  • Fragrance-free and dye-free
  • Contains Shea Butter to lock in hydration
  • Contains Allantoin for moisturization
  • Contains Glycerin for smooth skin
  • Silicone-free to prevent compatibility issues
  • Suitable for food facilities
  • Compatible with rubber production processes
  • Compatible with alcohol-based hand sanitizers
  • Proven glove compatibility
  • Chlorhexidine Gluconate (CHG) Compatible
  • Meets ECARF criteria for skin tolerability