Learn More
Amsbio LLC Human Stomach (Gastric) Cancer and Matched Metastasis FFPE Tissue Block, 2 blocks
Supplier:  Amsbio LLC AMS47007MP/DEL
Encompass
Human FFPE tissue blocks are processed from diseased tissues or clinically-unremarkable donor normal human tissue samples. Tissue is prepared by a certified medical pathologist, fixed in 10% neutral formalin for 24 hours and embedded in IHC-grade paraffin. The target FFPE specimen size is 0.5 x 0.5 x 0.5 cm, but it can vary significantly depending on the organ.
Catalog No. NC1684247
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.