Learn More
Seward™ Stomacher™ Blender Bag Clips
Catalog No. 13874850
Change view
Click to view available options
Packaging Quantity:
200/Pk.
Catalog No. Packaging Quantity
13-874-850 200/Pk.
1 options
Catalog No. 13-874-850 Supplier Seward™ Supplier No. 030021490
Add to Cart
Add to Cart
Seward™ Stomacher™ Bag Clips allow fast and easy securing for all 80 to 400 mL blender bags. Ideal for short term storage of prepared samples.
  • Eliminates leakage during storage or incubation
  • Reusable alternative to closure bags
  • Easily cleaned with most lab disinfectants (not autoclavable)
  • Suitable for all 80 to 400 mL blender bags
Specifications
Capacity (Metric) 80 to 400 mL
Material Plastic
Description Bag clips
For Use With (Application) Microbiology sample preparation
For Use With (Equipment) Blender bags
Packaging Quantity 200/Pk.
Packaging Type Box