Learn More
Aviva Systems Biology Corp STOML2 Antibody - C-terminal region (ARP61799_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61799_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STOML2. It was validated on Western Blot. . Alternate names: HSPC108, SLP-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-287-470