Learn More
Aviva Systems Biology Corp STOML3 Antibody - N-terminal region (ARP52851_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52851_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STOML3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STOML3. Alternate names: Epb7.2l, SROApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60850