Learn More
Cayman Chemical STOML3 Inhibitor OB-1, Purity: ≥98%, CAS: 300803-69-4, 10 mg
SDP
Catalog No. 502916074
Encompass_Preferred
An inhibitor of STOML3 oligomerization; inhibits oligomerization of mouse and human STOML3 in cell-free assays at 10 µM; reduces Piezo1 channel current amplitude in Neuro2a cells; inhibits tactile perception in wild-type mice and reduces tactile allodynia in a mouse model of streptozotocin-induced diabetic neuropathy
Catalog No. 50-291-6074 Supplier Cayman Chemical Supplier No. 3758410 MG
An inhibitor of STOML3 oligomerization; inhibits oligomerization of mouse and human STOML3 in cell-free assays at 10 µM; reduces Piezo1 channel current amplitude in Neuro2a cells; inhibits tactile perception in wild-type mice and reduces tactile allodynia in a mouse model of streptozotocin-induced diabetic neuropathy