Learn More
Cayman Chemical STOML3 Inhibitor OB-1, Purity: ≥98%, CAS: 300803-69-4, 25 mg
SDP
Catalog No. 502917612
Encompass_Preferred
An inhibitor of STOML3 oligomerization; inhibits oligomerization of mouse and human STOML3 in cell-free assays at 10 µM; reduces Piezo1 channel current amplitude in Neuro2a cells; inhibits tactile perception in wild-type mice and reduces tactile allodynia in a mouse model of streptozotocin-induced diabetic neuropathy
Catalog No. 50-291-7612 Supplier Cayman Chemical Supplier No. 3758425 MG
An inhibitor of STOML3 oligomerization; inhibits oligomerization of mouse and human STOML3 in cell-free assays at 10 µM; reduces Piezo1 channel current amplitude in Neuro2a cells; inhibits tactile perception in wild-type mice and reduces tactile allodynia in a mouse model of streptozotocin-induced diabetic neuropathy