Learn More
Clean Collect™ Blood Culture Kits
Description
Reducing blood culture contamination rates prevents unnecessary treatment of patients. Clean Collect kits can help to increase quality and save considerable resources.
- Prevents blood culture contamination using proprietary diversion method
- Average contamination rate of 0.5% compared to 3% national average
- Diversion method reduces contamination rates by an average of 74%
- Standardized kits promote consistency for long term improvement
- Reduces hospital costs considerably
- Ambient temperature (0-40°C) dry storage; 10 month shelf life
Specifications
Specifications
| Content And Storage | Diversion tube, standard needle hub, BD ChloraPrep™ swabsticks, sterile gloves, tourniquet, alcohol pads, bandage, instruction card, tracking decals; Store at ambient temperature (0-40°C) |
| Includes | Bactec,Versatrek; no needle |