Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vela Diagnostics Great Basin Stool Bacterial Pathogens Panel

Catalog No. 23052002
Change view
Click to view available options
Clia Complexity:
Moderate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
23-052-002 Moderate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-052-002 Supplier Vela Diagnostics Supplier No. 300780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Produces a definitive result in <2 hours, with less than 1 minute of hands-on time, detecting the most common bacterial causes of gastroenteritis directly from preserved patient stool specimen: C. jejuni and C. coli, Salmonella, Shigella, stx1, stx2, O157 E. coli serotype (if Shiga toxin pos)

An automated, qualitative DNA multiplex assay that quickly detects common bacterial pathogens directly from a patient’s preserved stool sample.

Specifications

Content And Storage 2°C to 8°C
Description Sample-to-result panel: Stool Bacterial Pathogens
Clia Complexity Moderate
Detectable Analytes Campylobacter (C. coli/C. jejuni), Salmonella, Shiga toxin 1 (stx1), Shiga toxin 2 (stx2), Escherichia coli serotype O157, Shigella
Sample Type Preserved Stool
Quantity 10 Assays
Test Time ≈90 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.