Learn More
Bioworld Stool DNA Isolation Kit, 50 Prep(s)
Catalog No. 50196940
Encompass_Preferred
For the rapid and simple purification of bacterial and host DNA from stool and fecal samples Simultaneous isolation of both host DNA and microbial DNA (universal protocol)Eliminates PCR inhibitors including humic acidHigh quality DNA for sensitive downstream applicationsFEATURES AND BENEFITSFast and easy processing - Rapid isolation of high-quality, ready-to-use DNA in 30 minutes.No organic extraction or alcohol precipitation- Phenol is not required for the isolationUniversal procedure - Simultaneous isolation of both host DNA and microbial DNAHigh yields - Consistent, high yieldsof inhibitor-free DNA up to 50 kb plusRemove all inhibitors- Eliminates all PCR inhibitors including humic acid
Catalog No. 50-196-940 Supplier Bioworld Supplier No. 107600831
For the rapid and simple purification of bacterial and host DNA from stool and fecal samples Simultaneous isolation of both host DNA and microbial DNA (universal protocol)Eliminates PCR inhibitors including humic acidHigh quality DNA for sensitive downstream applicationsFEATURES AND BENEFITSFast and easy processing - Rapid isolation of high-quality, ready-to-use DNA in 30 minutes.No organic extraction or alcohol precipitation- Phenol is not required for the isolationUniversal procedure - Simultaneous isolation of both host DNA and microbial DNAHigh yields - Consistent, high yieldsof inhibitor-free DNA up to 50 kb plusRemove all inhibitors- Eliminates all PCR inhibitors including humic acid