Learn More
Norgen Biotek Corp Stool Nucleic Acid Collection and Preservation Tubes, 50 Pack
Catalog No. NC1002341
Encompass_Preferred
Norgen’s Stool Nucleic Acid Collection and Preservation Tubes are for the rapid preservation of nucleic acids from fresh stool specimens. The Stool Nucleic Acid Collection and Preservation Tubes contain Norgen’s Stool Preservative in a liquid format. The Stool Preservative prevents the growth of Gram-negative and Gram-positive bacteria and fungi, and also inactivates viruses allowing the resulting non-infectious samples to be handled and shipped safely. In addition, the Stool Preservative eliminates the need to immediately process or freeze samples and allows the samples to be shipped to centralized testing facilities at ambient temperatures. Components of the Stool Preservative allow samples to be stored at room temperature for over 2 years for DNA and 7 days for RNA. To extend the stool RNA stability in the preservative, storage at -20°C or -70°C is recommended upon arrival at testing facilities until the RNA purification.
Catalog No. NC1002341 Supplier Norgen Biotek Corp Supplier No. 45660
Norgen’s Stool Nucleic Acid Collection and Preservation Tubes are for the rapid preservation of nucleic acids from fresh stool specimens. The Stool Nucleic Acid Collection and Preservation Tubes contain Norgen’s Stool Preservative in a liquid format. The Stool Preservative prevents the growth of Gram-negative and Gram-positive bacteria and fungi, and also inactivates viruses allowing the resulting non-infectious samples to be handled and shipped safely. In addition, the Stool Preservative eliminates the need to immediately process or freeze samples and allows the samples to be shipped to centralized testing facilities at ambient temperatures. Components of the Stool Preservative allow samples to be stored at room temperature for over 2 years for DNA and 7 days for RNA. To extend the stool RNA stability in the preservative, storage at -20°C or -70°C is recommended upon arrival at testing facilities until the RNA purification.