Learn More
Enterprise Technology Solutions Safco Economy Lab Stool, 18"x18"x16"-21", Black
SDP
Catalog No. 502173533
Encompass_Preferred
Multipurpose lab stool without back features long-wearing, black vinyl upholstery and a generously cushioned 13-1/2" diameter seat. Seat height adjusts pneumatically 5" from 16" to 21". Lab stool features a full 360-degree swivel seat, 2" swivel casters and 18" diameter, polished aluminum five-star base. Weight capacity is 250 lb. Stool meets or exceeds applicable ANSI/BIFMA standards.
Catalog No. 50-217-3533 Supplier Enterprise Technology Solutions Supplier No. SAF3434BL
Multipurpose lab stool without back features long-wearing, black vinyl upholstery and a generously cushioned 13-1/2" diameter seat. Seat height adjusts pneumatically 5" from 16" to 21". Lab stool features a full 360-degree swivel seat, 2" swivel casters and 18" diameter, polished aluminum five-star base. Weight capacity is 250 lb. Stool meets or exceeds applicable ANSI/BIFMA standards.