Learn More
BURKLE INC stop-it hose clamp, Easy-Click, Ø 0.79 in. (20 mm), blue
Catalog No. NC2187882
Encompass_Preferred
Stepless flow regulation: With the tubing clamp stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections. The tubing is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged.
This item is not returnable. View return policy
Catalog No. NC2187882 Supplier BURKLE INC Supplier No. 86190205
Stepless flow regulation: With the tubing clamp stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections. The tubing is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged.