Learn More
BURKLE INC stop-it hose clamp, Easy-Click, Ø 0.59 in. (15 mm), blue
Catalog No. 5000025815
Encompass_Preferred
Stepless flow regulation: With the tubing clamp stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections. The tubing is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged.
Catalog No. 50-000-25815 Supplier BURKLE INC Supplier No. 86190155
Stepless flow regulation: With the tubing clamp stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections. The tubing is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged.