Learn More
Alkali Scientific Stop-Pop™ Locking Clips, with Breakaway Lifting Tabs for 1.5mL Tubes, 100/cs
SDP
Catalog No. 502389359
Encompass_Preferred
Use Stop-Pops to prevent 1.5-2mL mini centrifuge tubes from popping open during heating / boiling. Stop-Pop Locking Clips contains a built-in lifting tab, making it easier to remove a hot mini centrifuge tube from a dry bath or boiling water bath.
Catalog No. 50-238-9359 Supplier Alkali Scientific Supplier No. MT2086
Use Stop-Pops to prevent 1.5-2mL mini centrifuge tubes from popping open during heating / boiling. Stop-Pop Locking Clips contains a built-in lifting tab, making it easier to remove a hot mini centrifuge tube from a dry bath or boiling water bath.