Learn More
Viro Labs STOP SOLUTION 100ML
Catalog No. NC9871413
Encompass
Ready-To-Use Stop Solution For One Component Tmb. Size: 100 Ml Application: Stop Enzyme Reaction And Enhance The Absorbance Of Tmb Oxidation Products. Storage: Store At Room Temperature (Up To 12 Months). For Best Result, Use This Stop Solution With Virolabs' Tmb Solution. Instruction: After 5-15 Min Incubation Of Tmb (E.G. 100 Ul) In The Microwells, Stop Solution (E.G. 100 Ul) Is Added To Quench The Enzyme Reaction And Convert The Tmb Oxidation Products From Blue To Yellow Color. Read Plates At 450 Nm.
Catalog No. NC9871413 Supplier Viro Labs Supplier No. RESTOP100
Ready-To-Use Stop Solution For One Component Tmb. Size: 100 Ml Application: Stop Enzyme Reaction And Enhance The Absorbance Of Tmb Oxidation Products. Storage: Store At Room Temperature (Up To 12 Months). For Best Result, Use This Stop Solution With Virolabs' Tmb Solution. Instruction: After 5-15 Min Incubation Of Tmb (E.G. 100 Ul) In The Microwells, Stop Solution (E.G. 100 Ul) Is Added To Quench The Enzyme Reaction And Convert The Tmb Oxidation Products From Blue To Yellow Color. Read Plates At 450 Nm.