Learn More
Enzo Life Sciences Stop solution 2, (10ml)
SDP
Catalog No. 502006811
Encompass_Preferred
Assay stop solution for HRP EIA/ELISA. Formulation: Liquid. 1N hydrochloric acid. Long Term Storage: Ambient.
Catalog No. 50-200-6811 Supplier Enzo Life Sciences Supplier No. ADI800377
Add to Cart
Add to Cart
Assay stop solution for HRP EIA/ELISA. Formulation: Liquid. 1N hydrochloric acid. Long Term Storage: Ambient.