Learn More
Enzo Life Sciences Stop solution 2, (50 ml) (50 ml)
SDP
Catalog No. 50658085
Encompass_Preferred
Acid stop solution (1N HCl) intended for use as an assay stop reagent for colorimetric HRP/TMB reactions. Each bottle is sufficient for use with 5x96 wells microtiter plates. Additional assay development reagents are available as companion products to our IMMUNOSET products. Shipping: Ambient. Long Term Storage: Ambient
Catalog No. 50-658-085 Supplier Enzo Life Sciences Supplier No. ADI801804
Acid stop solution (1N HCl) intended for use as an assay stop reagent for colorimetric HRP/TMB reactions. Each bottle is sufficient for use with 5x96 wells microtiter plates. Additional assay development reagents are available as companion products to our IMMUNOSET products. Shipping: Ambient. Long Term Storage: Ambient