Learn More
Ace Glass, Inc. Stopcock, 0-3mm, 90° arms, high vacuum, PTFE plug with knurled PTFE handle, completely bakeable
Supplier: Ace Glass, Inc. 819545
Stopcock, 0-3mm, 90° arms, high vacuum, PTFE plug with knurled PTFE handle, completely bakeable