Learn More
Ace Glass, Inc. Stopcock, 0-4mm, 90° arms, high vacuum, PTFE plug with knurled nylon handle
SDP
Catalog No. 50979724
Encompass_Preferred
Stopcock, 0-4mm, 90° arms, high vacuum, PTFE plug with knurled nylon handle
Catalog No. 50-979-724 Supplier Ace Glass, Inc. Supplier No. 819509
Stopcock, 0-4mm, 90° arms, high vacuum, PTFE plug with knurled nylon handle