Learn More
Ace Glass, Inc. Stopcock, 0-5mm, 90° arms, high vacuum, PTFE plug with hooded UHDPE handle
SDP
Catalog No. 50974441
Encompass_Preferred
Stopcock, 0-5mm, 90° arms, high vacuum, PTFE plug with hooded UHDPE handle
Catalog No. 50-974-441 Supplier Ace Glass, Inc. Supplier No. 8195240
Stopcock, 0-5mm, 90° arms, high vacuum, PTFE plug with hooded UHDPE handle