Learn More
BURKLE INC StopCock PA, electroconductive, cap nut 3/4" inner
Catalog No. 5000025837
Encompass_Preferred
Commercially available spigots made of the materials polyethylene or polypropylene cannot be used for solvents.
StopCock PA fits onto 3/4" outer threads and with the supplied adapter onto 3/4" inner threads and therefore onto nearly all commercially available canisters and barrels where the discharge thread is at the bottom.
The StopCock discharge cocks fit all containers with G 3/4" connection threads. Thread adapters facilitate connection of cocks to nearly all containers.
Catalog No. 50-000-25837 Supplier BURKLE INC Supplier No. 05251100
Commercially available spigots made of the materials polyethylene or polypropylene cannot be used for solvents.
StopCock PA fits onto 3/4" outer threads and with the supplied adapter onto 3/4" inner threads and therefore onto nearly all commercially available canisters and barrels where the discharge thread is at the bottom.
The StopCock discharge cocks fit all containers with G 3/4" connection threads. Thread adapters facilitate connection of cocks to nearly all containers.