Learn More
Chemglass Life Sciences Stopcock, Double Oblique, Hollow Glass Plug, High Vacuum, 2mm Bore, 14.5/50 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG47401
Encompass_Preferred
Hollow plug, double oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.
Catalog No. CG47401 Supplier Chemglass Life Sciences Supplier No. CG47401
Hollow plug, double oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.