Learn More
AVANTOR MASTERFLEX (VWR LLC) 3-Way PP Stopcock; 1/4- 5/16"" ID, 4 mm, 1/Pk
Catalog No. NC9408786
Encompass
Chemical and heat resistant PVDF stopcocks allow you to stop flow quickly. Easily connect hose barb connectors to flexible tubing for quick and easy installation.
Catalog No. NC9408786 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0622540
Chemical and heat resistant PVDF stopcocks allow you to stop flow quickly. Easily connect hose barb connectors to flexible tubing for quick and easy installation.