Learn More
Ace Glass, Inc. Vacuum take-off adapter, 24/40, 2mm double oblique bore, hose barbs for 5/16 or 3/8in ID tubing
SDP
Catalog No. 50979619
Encompass_Preferred
24/40 vacuum take-off adapter with 2mm double oblique bore, and hose barbs for 5/16 or 3/8 I.D. tubing. Used with 2000mL reaction kettle with sublimation apparatus accessories.
Catalog No. 50-979-619 Supplier Ace Glass, Inc. Supplier No. 802714
24/40 vacuum take-off adapter with 2mm double oblique bore, and hose barbs for 5/16 or 3/8 I.D. tubing. Used with 2000mL reaction kettle with sublimation apparatus accessories.