Learn More
Med Vet International Jorvet Discofix 3-Way Stop Cock, J0462
Catalog No. 502831802
Encompass_Preferred
Directional flow knob is labeled with "off." This is useful in recognizing flow direction. Sterile.
Catalog No. 50-283-1802 Supplier Med Vet International Supplier No. JOR462
Directional flow knob is labeled with "off." This is useful in recognizing flow direction. Sterile.