Learn More
Hach Company Stopcock Grease, 150 gram Tube
Supplier:  Hach Company 56275
Encompass
For lubricating ground glass joints, glass stopcocks and valves, Tensette Pipets, and for sealing desiccators. Lubricant has low vapor pressure and resists attack by acidic and alkaline solutions.
Catalog No. 50-199-4390