Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, High Vacuum, 4mm Bore, 17/40 Plug Size, 10mm OD Sidearms
SDP
Catalog No. 50873726
Encompass_Preferred
Solid glass high vacuum stopcock with theChemglass improved constant pressure retention device. This retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.
Catalog No. 50-873-726 Supplier Chemglass Life Sciences Supplier No. CG46202
Solid glass high vacuum stopcock with theChemglass improved constant pressure retention device. This retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.