Learn More
Warner Instruments™ Stopcock with Leur Connector
Catalog No. BTX640165 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
No. per Pack:
6 Pk.
Catalog No. No. per Pack
BTX640165 6 Pk.
1 options
Catalog No. BTX640165 Supplier Warner Instruments™ Supplier No. 640165
Stopcock with Luer connector is supplied as a spare part.
  • Supplied in pack of 6
Specifications
Type Stopcock
Plug Bore Conical Taper
Material Plastic
Autoclavable Not Autoclavable
No. per Pack 6 Pk.
Plug Material Polycarbonate
Outlet Port Tubulated
Connectors Luer