Learn More
Perkin Elmer US LLC Stopcock, Polycarb Replacement Valve for DigiFilter Manifold
Catalog No. 502103756
Encompass_Preferred
With the DigiFILTER Manifold, one can filter up to twelve DigiFILTERs connected at once. A vacuum pump draws suction from the manifold into clean DigiTUBE. Consistent sample filtrations at each position across the manifold. Multiple DigiFILTER Manifolds may be connected in a daisy chain.
Catalog No. 50-210-3756 Supplier Perkin Elmer US LLC Supplier No. N9308062
With the DigiFILTER Manifold, one can filter up to twelve DigiFILTERs connected at once. A vacuum pump draws suction from the manifold into clean DigiTUBE. Consistent sample filtrations at each position across the manifold. Multiple DigiFILTER Manifolds may be connected in a daisy chain.